accessibility menu, dialog, popup

UserName

ONO

CAS RN 446866-87-1

IUPAC Name: 3-(morpholin-4-yl)benzaldehyde
Synonyms: 3-(morpholin-4-yl)benzaldehyde
Molecular Weight (g/mol): 191.23
Molecular Formula: C11H13NO2
InChi Key: LQORKFSMUOSSQM-UHFFFAOYSA-N
SMILES: O=CC1=CC(=CC=C1)N1CCOCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results