accessibility menu, dialog, popup

UserName

CH3NNH3CCl

CAS RN 4472-45-1

IUPAC Name: 4-chloro-2,6-dimethylpyrimidine
Synonyms: 4-chloro-2,6-dimethylpyrimidine
Molecular Weight (g/mol): 142.59
Molecular Formula: C6H7ClN2
InChi Key: GSXFOGXQLRLSKK-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=NC(C)=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results