CAS RN 4488-22-6
IUPAC Name:
[1,1'-binaphthalene]-2,2'-diamine
Synonyms:
[1,1'-binaphthalene]-2,2'-diamine
Molecular Weight (g/mol):
284.36
Molecular Formula:
C20H16N2
InChi Key:
DDAPSNKEOHDLKB-UHFFFAOYSA-N
SMILES:
NC1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1N
1
–
1
of
1
results
1
(±)-1,1'-Bi(2-naphthylamine), 97%
CAS: 4488-22-6 Molecular Formula: C20H16N2 Molecular Weight (g/mol): 284.362 MDL Number: MFCD00145204 InChI Key: DDAPSNKEOHDLKB-UHFFFAOYSA-N Synonym: 1,1'-binaphthalene-2,2'-diamine,r-1,1'-binaphthalene-2,2'-diamine,s---1,1'-binaphthyl-2,2'-diamine,1,1'-binaphthyl-2,2'-diamine,s---2,2'-diamino-1,1'-binaphthalene,2,2'-diamino-1,1'-dinaphthyl,1,1'-bi 2-naphthylamine,1,1'-bi 2-naphthalenylamine,binam,r-+-2,2'-diamino-1,1'-binaphthalene PubChem CID: 20571 IUPAC Name: 1-(2-aminonaphthalen-1-yl)naphthalen-2-amine SMILES: C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)N)N