CAS RN 45102-52-1
CAS RN 45102-52-1
IUPAC Name:
2,2,3,3-tetrafluoropropyl 2-methylprop-2-enoate
Synonyms:
2,2,3,3-tetrafluoropropyl 2-methylprop-2-enoate
Molecular Weight (g/mol):
200.13
Molecular Formula:
C7H8F4O2
InChi Key:
RSVZYSKAPMBSMY-UHFFFAOYSA-N
SMILES:
CC(=C)C(=O)OCC(F)(F)C(F)F
1
–
1
of
1
results
1
2,2,3,3-Tetrafluoropropyl methacrylate, 97%, stab. with 50ppm 4-methoxyphenol, Thermo Scientific Chemicals
CAS: 45102-52-1 Molecular Formula: C7H8F4O2 Molecular Weight (g/mol): 200.133 MDL Number: MFCD00042381 InChI Key: RSVZYSKAPMBSMY-UHFFFAOYSA-N Synonym: 2,2,3,3-tetrafluoropropyl methacrylate,2-propenoic acid, 2-methyl-, 2,2,3,3-tetrafluoropropyl ester,methacrylic acid 2,2,3,3-tetrafluoropropyl ester,1h,1h,3h-tetrafluoropropyl methacrylate,2,2,3,3-tetrafluoropropyl methacrylate, stab. with 2,2'-methylenebis 6-tert-butyl-4-methyl-phenol,acmc-1ajz7,ksc489s8d,rsvzyskapmbsmy-uhfffaoysa,2,2,3,3-tetrafluoroprop-1-yl methacrylat PubChem CID: 123515 IUPAC Name: 2,2,3,3-tetrafluoropropyl 2-methylprop-2-enoate SMILES: CC(=C)C(=O)OCC(C(F)F)(F)F