accessibility menu, dialog, popup

UserName

CAS RN 4511-99-3

IUPAC Name: diphenyl-1,2,4-triazin-3-amine
Synonyms: diphenyl-1,2,4-triazin-3-amine
Molecular Weight (g/mol): 248.29
Molecular Formula: C15H12N4
InChi Key: NZRHOWNFGASHMN-UHFFFAOYSA-N
SMILES: NC1=NN=C(C2=CC=CC=C2)C(=N1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results