accessibility menu, dialog, popup

UserName

CAS RN 452-83-5

FH2NCl

CAS RN 452-83-5

IUPAC Name: 2-chloro-5-fluoroaniline
Synonyms: 2-chloro-5-fluoroaniline
Molecular Weight (g/mol): 145.56
Molecular Formula: C6H5ClFN
InChi Key: VWUFOZAFKYOZJB-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=CC(F)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results