accessibility menu, dialog, popup

UserName

CAS RN 455303-00-1

NOOOOOHOHOHOHH2OH2O

CAS RN 455303-00-1

IUPAC Name: 2-[(carboxylatomethyl)[(3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracen-2-yl)methyl]azaniumyl]acetate
Synonyms: 2-[(carboxylatomethyl)[(3,4-dihydroxy-9,10-dioxoanthracen-2-yl)methyl]ammonio]acetate
Molecular Weight (g/mol): 384.32
Molecular Formula: C19H14NO8
InChi Key: PWIGYBONXWGOQE-UHFFFAOYSA-M
SMILES: OC1=C(O)C2=C(C=C1C[NH+](CC([O-])=O)CC([O-])=O)C(=O)C1=CC=CC=C1C2=O
Color 
  • (2)
Grade 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)