accessibility menu, dialog, popup

UserName

CAS RN 4569-82-8

IUPAC Name: 2-phenyl-1,3-thiazolidine
Synonyms: 2-phenyl-1,3-thiazolidine
Molecular Weight (g/mol): 165.25
Molecular Formula: C9H11NS
InChi Key: NXOMVTLTYYYYPC-UHFFFAOYNA-N
SMILES: C1CSC(N1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results