accessibility menu, dialog, popup

UserName

NNHN

CAS RN 45887-08-9

IUPAC Name: 3-(1H-pyrazol-5-yl)pyridine
Synonyms: 3-(2H-pyrazol-3-yl)pyridine
Molecular Weight (g/mol): 145.17
Molecular Formula: C8H7N3
InChi Key: JJLMOUMOJSUSSX-UHFFFAOYSA-N
SMILES: N1N=CC=C1C1=CN=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results