accessibility menu, dialog, popup

UserName

CAS RN 4606-07-9

IUPAC Name: ethyl cyclopropanecarboxylate
Synonyms: cyclopropanecarboxylic acid ethyl ester
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: LDDOSDVZPSGLFZ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1CC1
Quantity 
  • (1)
  • (3)
  • (1)
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)