CAS RN 4619-20-9
IUPAC Name:
4-(4-methylphenyl)-4-oxobutanoic acid
Synonyms:
4-(4-methylphenyl)-4-oxobutanoic acid
Molecular Weight (g/mol):
192.21
Molecular Formula:
C11H12O3
InChi Key:
OEEUWZITKKSXAZ-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)C(=O)CCC(O)=O
1
–
1
of
1
results
1
3-(4-Methylbenzoyl)propionic acid, 98%
CAS: 4619-20-9 Molecular Formula: C11H12O3 Molecular Weight (g/mol): 192.214 MDL Number: MFCD00020541 InChI Key: OEEUWZITKKSXAZ-UHFFFAOYSA-N Synonym: 3-4-methylbenzoyl propionic acid,4-oxo-4-p-tolyl butanoic acid,4-4-methylphenyl-4-oxobutanoic acid,4-4-methylphenyl-4-oxobutyric acid,4-oxo-4-4-tolyl butyric acid,4-oxo-4-p-tolylbutyric acid,4-oxo-4-p-tolylbutanoic acid,3-4-methylbenzoyl-propionic acid,acmc-1aooh,benzenebutanoic acid, 4-methyl-gamma-oxo PubChem CID: 244162 IUPAC Name: 4-(4-methylphenyl)-4-oxobutanoic acid SMILES: CC1=CC=C(C=C1)C(=O)CCC(=O)O