CAS RN 4621-50-5
CAS RN 4621-50-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Phosphoric Acid Triheptyl Ester, TRC
CAS: 4621-50-5 Molecular Formula: C21H45O4P Molecular Weight (g/mol): 392.55 Synonym: Heptanol Phosphate,Heptyl Phosphate ((C7H15O)3PO) (7CI),Triheptyl Phosphate IUPAC Name: triheptyl phosphate SMILES: CCCCCCCOP(=O)(OCCCCCCC)OCCCCCCC