accessibility menu, dialog, popup

UserName

CAS RN 4641-57-0

IUPAC Name: 1-phenylpyrrolidin-2-one
Synonyms: 1-phenylpyrrolidin-2-one
Molecular Weight (g/mol): 161.2
Molecular Formula: C10H11NO
InChi Key: JMVIVASFFKKFQK-UHFFFAOYSA-N
SMILES: O=C1CCCN1C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results