accessibility menu, dialog, popup

UserName

CAS RN 465514-37-8

CH3NNFOH

CAS RN 465514-37-8

IUPAC Name: [1-(4-fluorophenyl)-5-methyl-1H-pyrazol-4-yl]methanol
Synonyms: [1-(4-fluorophenyl)-5-methylpyrazol-4-yl]methanol
Molecular Weight (g/mol): 206.22
Molecular Formula: C11H11FN2O
InChi Key: QJPGMVFNIWHOIY-UHFFFAOYSA-N
SMILES: CC1=C(CO)C=NN1C1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results