accessibility menu, dialog, popup

UserName

CAS RN 4656-81-9

H3CNCH3H3CCH3OClOOO

CAS RN 4656-81-9

IUPAC Name: tetrahexylazanium perchlorate
Synonyms: tetrahexylazanium perchlorate
Molecular Weight (g/mol): 454.13
Molecular Formula: C24H52ClNO4
InChi Key: RLCKPGXHGKSGOS-UHFFFAOYSA-M
SMILES: [O-][Cl](=O)(=O)=O.CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)