CAS RN 469-54-5
CAS RN 469-54-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Griseofulvic Acid, TRC
CAS: 469-54-5 Molecular Formula: C16 H15 Cl O6 Molecular Weight (g/mol): 338.74 Synonym: (1'S,6'R)-7-Chloro-4,6-dimethoxy-6'-methyl-spiro[benzofuran-2(3H),1'-[3]cyclohexane]-2',3,4'-trione,Spiro[benzofuran-2(3H),1'-cyclohexane]-2',3,4'-trione, 7-chloro-4,6-dimethoxy-6'-methyl-, trans- (8CI),(±)-Griseofulvic acid,Griseofulvic acid IUPAC Name: (2S,5'R)-7-chloro-4,6-dimethoxy-5'-methylspiro[1-benzofuran-2,4'-cyclohexane]-1',3,3'-trione SMILES: COc1cc(OC)c2C(=O)[C@]3(Oc2c1Cl)[C@H](C)CC(=O)CC3=O