accessibility menu, dialog, popup

UserName

H3CCH3NN

CAS RN 4733-39-5

IUPAC Name: 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline
Synonyms: 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline
Molecular Weight (g/mol): 360.46
Molecular Formula: C26H20N2
InChi Key: STTGYIUESPWXOW-UHFFFAOYSA-N
SMILES: CC1=CC(C2=CC=CC=C2)=C2C=CC3=C(C=C(C)N=C3C2=N1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (3)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (3)
  • (4)
  • (3)
  • (3)
  • (1)
  • (2)