accessibility menu, dialog, popup

UserName

CAS RN 47375-34-8

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-[4-(tert-butoxy)phenyl]propanoic acid
Synonyms: 3-[4-(tert-butoxy)phenyl]-2-[(tert-butoxycarbonyl)amino]propanoic acid
Molecular Weight (g/mol): 337.42
Molecular Formula: C18H27NO5
InChi Key: ZEQLLMOXFVKKCN-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC1=CC=C(OC(C)(C)C)C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results