CAS RN 476014-76-3
CAS RN 476014-76-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Hydroxy-2-piperidinone, TRC
CAS: 476014-76-3 Molecular Formula: C5H9NO2 Molecular Weight (g/mol): 115.13 Synonym: 2-Piperidinone, 4-hydroxy-,4-Hydroxy-2-piperidinone,4-Hydroxypiperidin-2-one IUPAC Name: 4-hydroxypiperidin-2-one SMILES: OC1CCNC(=O)C1