accessibility menu, dialog, popup

UserName

CAS RN 4769-96-4

ONOHN

CAS RN 4769-96-4

IUPAC Name: 6-nitro-1H-indole
Synonyms: 6-nitro-1H-indole
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: PSWCIARYGITEOY-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2C=CNC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results