accessibility menu, dialog, popup

UserName

CAS RN 477575-56-7

H3CNHONNCH3SOOClClHNO(E/Z)

CAS RN 477575-56-7

IUPAC Name: 5-[(2,6-dichlorophenyl)methanesulfonyl]-3-[(3,5-dimethyl-4-{2-[(pyrrolidin-1-yl)methyl]pyrrolidine-1-carbonyl}-1H-pyrrol-2-yl)methylidene]-2,3-dihydro-1H-indol-2-one
Synonyms: 5-[(2,6-dichlorophenyl)methanesulfonyl]-3-({3,5-dimethyl-4-[2-(pyrrolidin-1-ylmethyl)pyrrolidine-1-carbonyl]-1H-pyrrol-2-yl}methylidene)-1H-indol-2-one
Molecular Weight (g/mol): 641.61
Molecular Formula: C32H34Cl2N4O4S
InChi Key: OYONTEXKYJZFHA-UHFFFAOYNA-N
SMILES: CC1=C(C(=O)N2CCCC2CN2CCCC2)C(C)=C(N1)C=C1C(=O)NC2=C1C=C(C=C2)S(=O)(=O)CC1=C(Cl)C=CC=C1Cl
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results