CAS RN 4787-77-3
IUPAC Name:
2-(pyrrolidin-1-yl)phenol
Synonyms:
2-(pyrrolidin-1-yl)phenol
Molecular Weight (g/mol):
163.22
Molecular Formula:
C10H13NO
InChi Key:
ZGSBDRFDXWRZAE-UHFFFAOYSA-N
SMILES:
OC1=CC=CC=C1N1CCCC1
1
–
1
of
1
results
1
2-(1-Pyrrolidinyl)phenol, 98%
CAS: 4787-77-3 Molecular Formula: C10H13NO Molecular Weight (g/mol): 163.22 MDL Number: MFCD00051659 InChI Key: ZGSBDRFDXWRZAE-UHFFFAOYSA-N Synonym: 2-pyrrolidin-1-yl phenol,2-pyrrolidinophenol,2-1-pyrrolidino phenol,2-1-pyrrolidinyl phenol,2-tetrahydro-1h-pyrrol-1-ylphenol,2-pyrrolidinylphenol,2-pyrrolizinophenol,2-pyrrolidin-1-yl-phenol,phenol, 2-1-pyrrolidinyl,1-2-hydroxyphenyl pyrrolidine PubChem CID: 2734845 IUPAC Name: 2-pyrrolidin-1-ylphenol SMILES: OC1=CC=CC=C1N1CCCC1