accessibility menu, dialog, popup

UserName

N(R)NO(S)(R)ONHHOHOO

CAS RN 4836-13-9

IUPAC Name: N-{1-[(2S,4R,5S)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-2-oxo-1,2-dihydropyrimidin-4-yl}benzamide
Synonyms: N-{1-[(2S,4R,5S)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-2-oxopyrimidin-4-yl}benzamide
Molecular Weight (g/mol): 331.33
Molecular Formula: C16H17N3O5
InChi Key: MPSJHJFNKMUKCN-DYEKYZERSA-N
SMILES: OC[C@@H]1O[C@@H](C[C@H]1O)N1C=CC(NC(=O)C2=CC=CC=C2)=NC1=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results