accessibility menu, dialog, popup

UserName

H3CCl

CAS RN 4860-03-1

IUPAC Name: 1-chlorohexadecane
Synonyms: 1-chlorohexadecane
Molecular Weight (g/mol): 260.89
Molecular Formula: C16H33Cl
InChi Key: CLWAXFZCVYJLLM-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCl
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)