accessibility menu, dialog, popup

UserName

BrOHN

CAS RN 4876-10-2

IUPAC Name: 4-(bromomethyl)-1,2-dihydroquinolin-2-one
Synonyms: 4-(bromomethyl)-1H-quinolin-2-one
Molecular Weight (g/mol): 238.08
Molecular Formula: C10H8BrNO
InChi Key: BBAHJCUCNVVEQU-UHFFFAOYSA-N
SMILES: BrCC1=CC(=O)NC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)