accessibility menu, dialog, popup

UserName

CH3N

CAS RN 491-35-0

IUPAC Name: 4-methylquinoline
Synonyms: 4-methylquinoline 4-lepidine cincholepidine lepidine lepidin gamma-methylquinoline
Molecular Weight (g/mol): 143.19
Molecular Formula: C10H9N
InChi Key: MUDSDYNRBDKLGK-UHFFFAOYSA-N
SMILES: CC1=C2C=CC=CC2=NC=C1
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)