accessibility menu, dialog, popup

UserName

CH3O(E)

CAS RN 495-45-4

IUPAC Name: (2E)-1,3-diphenylbut-2-en-1-one
Synonyms: 2-buten-1-one, 1,3-diphenyl-
Molecular Weight (g/mol): 222.29
Molecular Formula: C16H14O
InChi Key: PLELHVCQAULGBH-OUKQBFOZSA-N
SMILES: C\C(=C/C(=O)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results