accessibility menu, dialog, popup

UserName

CAS RN 497-76-7

HOOOOHOHHOHO

CAS RN 497-76-7

IUPAC Name: 2-(hydroxymethyl)-6-(4-hydroxyphenoxy)oxane-3,4,5-triol
Synonyms: arbutin ursin
Molecular Weight (g/mol): 272.25
Molecular Formula: C12H16O7
InChi Key: BJRNKVDFDLYUGJ-UHFFFAOYNA-N
SMILES: OCC1OC(OC2=CC=C(O)C=C2)C(O)C(O)C1O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)