accessibility menu, dialog, popup

UserName

CAS RN 499770-91-1

IUPAC Name: 1-(3,4-dihydro-2H-1,5-benzodioxepin-6-yl)methanamine
Synonyms: 1-(3,4-dihydro-2H-1,5-benzodioxepin-6-yl)methanamine
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: ZXJOKQNPRXXPJT-UHFFFAOYSA-N
SMILES: NCC1=C2OCCCOC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results