accessibility menu, dialog, popup

UserName

CAS RN 499770-96-6

OOBr

CAS RN 499770-96-6

IUPAC Name: 6-(bromomethyl)-3,4-dihydro-2H-1,5-benzodioxepine
Synonyms: 6-(bromomethyl)-3,4-dihydro-2H-1,5-benzodioxepine
Molecular Weight (g/mol): 243.1
Molecular Formula: C10H11BrO2
InChi Key: QDRNTKFXSFDJCH-UHFFFAOYSA-N
SMILES: BrCC1=C2OCCCOC2=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results