CAS RN 500-67-4
CAS RN 500-67-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-Heptylresorcinol, TRC
CAS: 500-67-4 Molecular Formula: C13 H20 O2 Molecular Weight (g/mol): 208.3 Synonym: Spherophorol,5-Heptyl-1,3-benzenediol,3,5-Dihydroxy-1-heptylbenzene IUPAC Name: 5-heptylbenzene-1,3-diol SMILES: CCCCCCCc1cc(O)cc(O)c1