CAS RN 501418-06-0
CAS RN 501418-06-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1H,1aH,6H,6aH-cyclopropa[a]indene-1-carboxylic acid, TRC
CAS: 501418-06-0 Molecular Formula: C11H10O2 Molecular Weight (g/mol): 174.2 IUPAC Name: 1,1a,6,6a-tetrahydrocyclopropa[a]indene-1-carboxylic acid SMILES: OC(=O)C1C2Cc3ccccc3C12