accessibility menu, dialog, popup

UserName

O

CAS RN 502-49-8

IUPAC Name: cyclooctanone
Synonyms: cyclooctanone
Molecular Weight (g/mol): 126.2
Molecular Formula: C8H14O
InChi Key: IIRFCWANHMSDCG-UHFFFAOYSA-N
SMILES: O=C1CCCCCCC1
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (3)
  • (3)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)