accessibility menu, dialog, popup

UserName

H3CNOO

CAS RN 5022-29-7

IUPAC Name: 2-ethyl-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-ethylisoindole-1,3-dione
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: JZDSOQSUCWVBMV-UHFFFAOYSA-N
SMILES: CCN1C(=O)C2=CC=CC=C2C1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results