CAS RN 503-38-8
IUPAC Name:
trichloromethyl carbonochloridate
Synonyms:
diphosgene
Molecular Weight (g/mol):
197.82
Molecular Formula:
C2Cl4O2
InChi Key:
HCUYBXPSSCRKRF-UHFFFAOYSA-N
SMILES:
ClC(=O)OC(Cl)(Cl)Cl
1
–
2
of
2
results
1
Trichloromethyl chloroformate, 98%
CAS: 503-38-8 Molecular Formula: C2Cl4O2 Molecular Weight (g/mol): 197.82 MDL Number: MFCD00015553 InChI Key: HCUYBXPSSCRKRF-UHFFFAOYSA-N Synonym: diphosgene,trichloromethyl chloroformate,diphosgen,difosgen czech,perchloromethyl formate,unii-po4q4r80lv,hsdb 371,carbonochloridic acid, trichloromethyl ester,methanol, trichloro-, chloroformate,chloroformic acid trichloromethyl ester PubChem CID: 10426 IUPAC Name: trichloromethyl carbonochloridate SMILES: ClC(=O)OC(Cl)(Cl)Cl
2
Trichloromethyl Chloroformate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.