CAS RN 5035-82-5
CAS RN 5035-82-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methyl 2-Amino-3,4,5-trimethoxybenzoate, TRC
CAS: 5035-82-5 Molecular Formula: C11H15NO5 Molecular Weight (g/mol): 241.24 IUPAC Name: methyl 2-amino-3,4,5-trimethoxybenzoate SMILES: COC(=O)c1cc(OC)c(OC)c(OC)c1N