accessibility menu, dialog, popup

UserName

BrNH2N

CAS RN 50358-42-4

IUPAC Name: 5-bromoquinolin-6-amine
Synonyms: 5-bromoquinolin-6-amine
Molecular Weight (g/mol): 223.07
Molecular Formula: C9H7BrN2
InChi Key: MODLGTLYXJGDCH-UHFFFAOYSA-N
SMILES: NC1=C(Br)C2=CC=CN=C2C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results