accessibility menu, dialog, popup

UserName

ClH3CCH2OONCH3CH3H3C

CAS RN 5039-78-1

IUPAC Name: trimethyl({2-[(2-methylprop-2-enoyl)oxy]ethyl})azanium chloride
Synonyms: polyquaternium-37
Molecular Weight (g/mol): 207.7
Molecular Formula: C9H18ClNO2
InChi Key: RRHXZLALVWBDKH-UHFFFAOYSA-M
SMILES: [Cl-].CC(=C)C(=O)OCC[N+](C)(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results