accessibility menu, dialog, popup

UserName

CAS RN 50533-97-6

H3CNCH3NH

CAS RN 50533-97-6

IUPAC Name: N,N-dimethylpiperidin-4-amine
Synonyms: N,N-dimethylpiperidin-4-amine
Molecular Weight (g/mol): 128.22
Molecular Formula: C7H16N2
InChi Key: YFJAIURZMRJPDB-UHFFFAOYSA-N
SMILES: CN(C)C1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results