accessibility menu, dialog, popup

UserName

CAS RN 506-96-7

CH3BrO

CAS RN 506-96-7

IUPAC Name: acetyl bromide
Synonyms: acetyl bromide
Molecular Weight (g/mol): 122.95
Molecular Formula: C2H3BrO
InChi Key: FXXACINHVKSMDR-UHFFFAOYSA-N
SMILES: CC(Br)=O
Boiling Point 
  • (5)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)