accessibility menu, dialog, popup

UserName

H3COON

CAS RN 50743-18-5

IUPAC Name: 6-methyl-4-oxo-4H-chromene-3-carbonitrile
Synonyms: 6-methyl-4-oxochromene-3-carbonitrile
Molecular Weight (g/mol): 185.18
Molecular Formula: C11H7NO2
InChi Key: FNRLRLMPBIRUSQ-UHFFFAOYSA-N
SMILES: CC1=CC=C2OC=C(C#N)C(=O)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results