accessibility menu, dialog, popup

UserName

OHIO

CAS RN 50765-11-2

IUPAC Name: 5-hydroxy-2-iodobenzaldehyde
Synonyms: 5-hydroxy-2-iodobenzaldehyde
Molecular Weight (g/mol): 248.02
Molecular Formula: C7H5IO2
InChi Key: JRJRYFRHGBHRHI-UHFFFAOYSA-N
SMILES: OC1=CC=C(I)C(C=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results