accessibility menu, dialog, popup

UserName

CH3OBr

CAS RN 51012-65-8

IUPAC Name: 2-bromo-1-(2-methylphenyl)ethan-1-one
Synonyms: 2-bromo-1-(2-methylphenyl)ethanone
Molecular Weight (g/mol): 213.07
Molecular Formula: C9H9BrO
InChi Key: XMGAXELQRATLJP-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1C(=O)CBr
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results