accessibility menu, dialog, popup

UserName

H3COOOOH

CAS RN 51019-43-3

IUPAC Name: 2-(acetyloxy)-2-phenylacetic acid
Synonyms: (acetyloxy)(phenyl)acetic acid
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: OBCUSTCTKLTMBX-UHFFFAOYNA-N
SMILES: CC(=O)OC(C(O)=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results