CAS RN 51145-57-4
IUPAC Name:
ethyl (2E)-2-[(dimethylamino)methylidene]-3-oxobutanoate
Synonyms:
ethyl (2E)-2-[(dimethylamino)methylidene]-3-oxobutanoate
Molecular Weight (g/mol):
185.22
Molecular Formula:
C9H15NO3
InChi Key:
LQSOVGAUOHMPLK-SOFGYWHQSA-N
SMILES:
CCOC(=O)C(=C\N(C)C)\C(C)=O
1
–
4
of
4
results
1
Ethyl 2-acetyl-3-(dimethylamino)acrylate, 95%, Thermo Scientific Chemicals
CAS: 51145-57-4 Molecular Formula: C9H15NO3 Molecular Weight (g/mol): 185.223 MDL Number: MFCD00087563 InChI Key: LQSOVGAUOHMPLK-SOFGYWHQSA-N PubChem CID: 2763877 IUPAC Name: ethyl (2E)-2-(dimethylaminomethylidene)-3-oxobutanoate SMILES: CCOC(=O)C(=CN(C)C)C(=O)C
2
Ethyl 2-Dimethylaminomethylene-3-oxobutanoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Ethyl 2-Dimethylaminomethylene-3-oxobutanoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Ethyl 2-Dimethylaminomethylene-3-oxobutanoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.