accessibility menu, dialog, popup

UserName

BrBrH2C

CAS RN 513-31-5

IUPAC Name: 2,3-dibromoprop-1-ene
Synonyms: 2,3-dibromo-1-propene
Molecular Weight (g/mol): 199.87
Molecular Formula: C3H4Br2
InChi Key: YMFWYDYJHRGGPF-UHFFFAOYSA-N
SMILES: BrCC(Br)=C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)