CAS RN 51322-69-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(4,5-Dihydro-1H-imidazol-2-yl)-2,1,3-benzothiadiazol-4-amine, TRC
CAS: 51322-69-1 Molecular Formula: C9H9N5S Molecular Weight (g/mol): 219.27 Synonym: 2,1,3-Benzothiadiazol-4-amine, N-(4,5-dihydro-1H-imidazol-2-yl)-,N-(4,5-Dihydro-1H-imidazol-2-yl)-2,1,3-benzothiadiazol-4-amine,Deschlorotizanidine,Deschloro tizanidine,Dechloro tizanidine SMILES: C1CN=C(Nc2cccc3nsnc23)N1