accessibility menu, dialog, popup

UserName

NONBr

CAS RN 51376-06-8

IUPAC Name: 5-bromo-2,1,3-benzoxadiazole
Synonyms: 5-bromo-2,1,3-benzoxadiazole
Molecular Weight (g/mol): 199.01
Molecular Formula: C6H3BrN2O
InChi Key: ZWDFFESFCIACQC-UHFFFAOYSA-N
SMILES: BrC1=CC2=NON=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)