accessibility menu, dialog, popup

UserName

ClOCl

CAS RN 51512-09-5

IUPAC Name: 2-(2-chlorophenyl)acetyl chloride
Synonyms: 2-(2-chlorophenyl)acetyl chloride
Molecular Weight (g/mol): 189.04
Molecular Formula: C8H6Cl2O
InChi Key: WIHSAOYVGKVRJX-UHFFFAOYSA-N
SMILES: ClC(=O)CC1=CC=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results