accessibility menu, dialog, popup

UserName

Cl(R)O

CAS RN 51594-55-9

IUPAC Name: (2R)-2-(chloromethyl)oxirane
Synonyms: (-)-epichlorohydrin (R)-(-)-epichlorohydrin (R)-epichlorohydrin
Molecular Weight (g/mol): 92.52
Molecular Formula: C3H5ClO
InChi Key: BRLQWZUYTZBJKN-VKHMYHEASA-N
SMILES: ClC[C@H]1CO1
Quantity 
  • (2)
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)